siRNA Details
Gene Symbol: Ccna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007628.3 | NP_031654.2 | 5, 6 | 877 | ||
GenBank mRNA | AK077114.1 | - | 5, 6 | 883 | ||
BC120518.1 | - | 4, 5 | 792 | 40129441 | ||
BC125436.1 | - | 4, 5 | 792 | 40129951 | ||
X84311.1 | - | 5, 6 | 876 |

More Information
Other Species Targeted

Gene Ontology Categories:
