siRNA Details
Gene Symbol: Ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007631.2 | NP_031657.1 | 2 | 601 | ||
GenBank mRNA | AK005352.1 | - | 2 | 295 | ||
AK017834.1 | - | 2 | 565 | |||
AK053348.1 | - | 2 | 509 | |||
AK149905.1 | - | 2 | 508 | |||
AK164997.1 | - | 2 | 546 | |||
AK170833.1 | - | 2 | 482 | |||
BC044841.1 | - | 2 | 504 | 3155470 | ||
BC060172.1 | - | 2 | 496 | |||
EU600170.1 | - | 2 | 601 | |||
M64403.1 | - | 2 | 556 | |||
S78355.1 | - | 2 | 506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
