siRNA Details
Gene Symbol: Ccnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009833.1 | NP_033963.1 | 2 | 206 | ||
GenBank mRNA | AF087662.1 | - | 2 | 206 | ||
AF095640.1 | - | 2 | 206 | |||
AF109179.1 | - | 2 | 206 | |||
AF113951.1 | - | 2 | 206 | |||
AK083037.1 | - | 2 | 250 | |||
AK133168.1 | - | 906 | ||||
BC131685.1 | - | 2 | 206 | 40111509 | ||
BC131686.1 | - | 2 | 206 |
