siRNA Details
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007637.2 | NP_031663.1 | 11 | 1613 | ||
GenBank mRNA | AK077753.1 | - | 11 | 1572 | ||
AK088184.1 | - | 11 | 1547 | |||
AK129056.1 | - | 11 | 1519 | |||
AK133857.1 | - | 11 | 1571 | |||
AK136493.1 | - | 11 | 1565 | |||
AK145445.1 | - | 11 | 1582 | |||
AK146405.1 | - | 11 | 1585 | |||
AK146425.1 | - | 11 | 1565 | |||
AK150390.1 | - | 11 | 1532 | |||
AK153911.1 | - | 11 | 1589 | |||
AK167895.1 | - | 11 | 1554 | |||
AK213471.1 | - | 3 | 287 | |||
AK216446.1 | - | 3 | 225 | |||
AK219685.1 | - | 2 | 209 | |||
BC092233.1 | - | 11 | 1583 | |||
BC094427.1 | - | 11 | 1560 | 30537966 | ||
Z31555.1 | - | 11 | 1613 |

More Information
Other Species Targeted

Gene Ontology Categories:
