siRNA Details
Gene Symbol: Cct6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291242.1 | NP_001278171.1 | 11 | 1499 | ||
NM_009839.3 | NP_033969.2 | 11 | 1430 | |||
GenBank mRNA | AK076639.1 | - | 11 | 1500 | ||
BC100457.1 | - | 11 | 1397 | 30918851 | ||
CF551313.1 | - | 3 | 316 | |||
Z50192.1 | - | 11 | 1415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
