siRNA Details
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007641.5 | NP_031667.1 | 5 | 497 | ||
GenBank mRNA | AK017903.1 | - | 5 | 498 | ||
AK041428.1 | - | 5 | 496 | |||
AK080040.1 | - | 5 | 463 | |||
AK080148.1 | - | 495 | ||||
AK137469.1 | - | 5 | 486 | |||
AK143824.1 | - | 4 | 2375 | |||
AK156033.1 | - | 496 | ||||
AK166571.1 | - | 5 | 591 | |||
AK166659.1 | - | 5 | 591 | |||
AK171631.1 | - | 5 | 526 | |||
AK171638.1 | - | 5 | 449 | |||
AK171659.1 | - | 5 | 482 | |||
BC028322.1 | - | 5 | 478 | 4503841 | ||
CT010253.1 | - | 4 | 378 | |||
M62541.1 | - | 555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
