siRNA Details
Gene Symbol: Scarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007644.3 | NP_031670.1 | 3 | 631 | ||
GenBank mRNA | AB008553.1 | - | 3 | 536 | ||
AK011123.1 | - | 3 | 561 | |||
AK083038.1 | - | 3 | 632 | |||
AK144235.1 | - | 3 | 663 | |||
AK149102.1 | - | 577 | ||||
BC029073.1 | - | 3 | 539 | 3962669 |
