siRNA Details
Gene Symbol: Cd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290802.1 | NP_001277731.1 | 3 | 290 | ||
NM_001290804.1 | NP_001277733.1 | 2 | 215 | |||
NM_007645.4 | NP_031671.1 | 2 | 239 | |||
GenBank mRNA | AK154250.1 | - | 2 | 276 | ||
AK154466.1 | - | 3 | 260 | |||
AK155663.1 | - | 2 | 216 | |||
AK155966.1 | - | 2 | 204 | |||
BC004610.1 | - | 2 | 196 | |||
BC019402.1 | - | 2 | 276 | 4013070 | ||
BC031724.1 | - | 2 | 196 | |||
BE849269.1 | - | 2 | 157 | |||
CT010217.1 | - | 2 | 128 | |||
CX226822.1 | - | 3 | 291 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017124, XM_008759326, XM_006228981, BC059144, X53517, and others.
Gene Ontology Categories:
