siRNA Details
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009849.2 | NP_033979.2 | 8 | 1372 | ||
GenBank mRNA | AF042811.1 | - | 8 | 1296 | ||
AK002553.1 | - | 8 | 1344 | |||
AK172281.1 | - | 8 | 1320 | |||
AY376711.1 | - | 8 | 1305 | |||
BC011241.1 | - | 8 | 1304 | 3582550 | ||
U91511.1 | - | 6 | 1259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
