siRNA Details
Gene Symbol: Cd3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007648.4 | NP_031674.1 | 3, 4 | 149 | ||
GenBank mRNA | AK041607.1 | - | 3, 4 | 152 | ||
BC028636.1 | - | 2, 3 | 132 | |||
BC098236.1 | - | 2, 3 | 127 | |||
BC145926.1 | - | 3, 4 | 149 | 40131229 | ||
BC145954.1 | - | 3, 4 | 149 | 40131257 | ||
EF474454.2 | - | 2, 3 | 71 | |||
GU456631.1 | - | 2, 3 | 71 | |||
J02990.1 | - | 3, 4 | 150 |
