siRNA Details
Gene Symbol: Cd3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009850.2 | NP_033980.1 | 3 | 284 | ||
GenBank mRNA | AK134049.1 | - | 3 | 217 | ||
AK153980.1 | - | 3 | 216 | |||
AK153987.1 | - | 3 | 203 | |||
BC027528.1 | - | 3 | 215 | 1494674 | ||
BY224047.1 | - | 2 | 260 | |||
EF474453.1 | - | 3 | 173 | |||
GU456630.1 | - | 3 | 173 | |||
M58149.1 | - | 3 | 190 | |||
Y00635.1 | - | 3 | 312 |
