siRNA Details
Gene Symbol: Cd247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113393.2 | NP_001106864.2 | 8 | 621 | ||
NM_031162.4 | NP_112439.1 | 8 | 624 | |||
GenBank mRNA | AK016078.1 | - | 133 | |||
L03353.1 | - | 8 | 537 | |||
M33158.1 | - | 8 | 614 | |||
S51932.1 | - | 8 | 537 | |||
U56258.1 | - | 8 | 540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
