siRNA Details
Gene Symbol: Cd69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033122.3 | NP_001028294.1 | 3 | 369 | ||
GenBank mRNA | AK017979.1 | - | 3 | 370 | ||
AK151076.1 | - | 3 | 370 | |||
AK153224.1 | - | 3 | 370 | |||
BC106997.2 | - | 3 | 354 | |||
BC128272.1 | - | 3 | 354 | 40081252 | ||
BC128273.1 | - | 3 | 354 | 40081255 | ||
BC139135.1 | - | 3 | 331 | 8862157 | ||
BC139136.1 | - | 3 | 331 | 8862158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
