siRNA Details
Gene Symbol: Cd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009854.2 | NP_033984.1 | 2 | 275 | ||
GenBank mRNA | AK136444.1 | - | 2 | 229 | ||
AK154339.1 | - | 2 | 230 | |||
BC024376.1 | - | 2 | 216 | 4985794 | ||
CT010368.1 | - | 2 | 162 | |||
D10329.1 | - | 2 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
