siRNA Details
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110320.1 | NP_001103790.1 | 6 | 917 | ||
NM_001110321.1 | NP_001103791.1 | 5 | 845 | |||
NM_001110322.1 | NP_001103792.1 | 4 | 755 | |||
NM_007654.2 | NP_031680.2 | 6 | 917 | |||
GenBank mRNA | AA203771.1 | - | 1 | 91 | ||
AK134132.1 | - | 6 | 920 | |||
AK144191.1 | - | 5 | 848 | |||
AK151094.1 | - | 757 | ||||
J04170.1 | - | 6 | 854 | |||
S40777.1 | - | 6 | 850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
