siRNA Details
Gene Symbol: Cd8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081110.2 | NP_001074579.1 | 4 | 800 | ||
GenBank mRNA | AK088128.1 | - | 4 | 680 | ||
AK088214.1 | - | 4 | 680 | |||
AK088447.1 | - | 3 | 1185 | |||
AK137937.1 | - | 4 | 651 | |||
BC030679.1 | - | 4 | 664 | 1247019 | ||
M12052.1 | - | 4 | 800 | |||
M12825.1 | - | 4 | 686 | |||
U34881.1 | - | 4 | 674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
