siRNA Details
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007657.3 | NP_031683.1 | 2 | 259 | ||
GenBank mRNA | AK002251.1 | - | 2 | 265 | ||
AK012793.1 | - | 2 | 265 | |||
AK151443.1 | - | 2 | 225 | |||
AK151619.1 | - | 2 | 212 | |||
AK170310.1 | - | 2 | 212 | |||
BC070474.1 | - | 2 | 259 | 30634238 | ||
L08115.1 | - | 2 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
