siRNA Details
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243769.1 | NP_001230698.1 | 3 | 300 | ||
NM_009861.3 | NP_033991.1 | 3 | 300 | |||
GenBank mRNA | AK003098.1 | - | 3 | 221 | ||
AK051543.1 | - | 3 | 221 | |||
AK075567.1 | - | 3 | 219 | |||
AK081384.1 | - | 1 | 2379 | |||
AK144216.1 | - | 3 | 223 | |||
AK145659.1 | - | 2 | 64 | |||
AK150563.1 | - | 3 | 234 | |||
AK151087.1 | - | 3 | 245 | |||
AK151726.1 | - | 3 | 222 | |||
AK153564.1 | - | 3 | 244 | |||
AK154870.1 | - | 3 | 219 | |||
AK159470.1 | - | 3 | 262 | |||
AK166281.1 | - | 3 | 219 | |||
AK167195.1 | - | 3 | 208 | |||
AK167400.1 | - | 3 | 213 | |||
AK167609.1 | - | 3 | 218 | |||
AK168013.1 | - | 2 | 213 | |||
AK168076.1 | - | 3 | 213 | |||
AK168089.1 | - | 2 | 213 | |||
AK168276.1 | - | 3 | 213 | |||
AK168503.1 | - | 3 | 234 | |||
AK168758.1 | - | 3 | 253 | |||
AK168820.1 | - | 3 | 226 | |||
AK169122.1 | - | 3 | 234 | |||
AK169232.1 | - | 3 | 213 | |||
AK169805.1 | - | 3 | 238 | |||
AK213611.1 | - | 2 | 74 | |||
BC064792.1 | - | 3 | 206 | 30105331 | ||
CD551112.1 | - | 3 | 287 | |||
L11318.1 | - | 2 | 136 | |||
L78075.1 | - | 2 | 140 | |||
M79301.1 | - | 2 | 94 | |||
U37720.1 | - | 3 | 208 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_171994, XM_008764286, XM_008764287, FQ231846, FQ232179, and others.
Gene Ontology Categories:
