siRNA Details
Gene Symbol: Arhgap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020260.2 | NP_064656.2 | 12 | 3405 | ||
GenBank mRNA | AK129309.1 | - | 3 | 1830 | ||
AK149975.1 | - | 12 | 3241 | |||
AK150109.1 | - | 12 | 3241 | |||
AK155729.1 | - | 12 | 3225 | |||
AK170685.1 | - | 12 | 3225 | |||
BC138870.1 | - | 12 | 3210 | 8861892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
