siRNA Details
Gene Symbol: Cdh15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007662.2 | NP_031688.2 | 12 | 2065 | ||
GenBank mRNA | BC157978.1 | - | 12 | 1982 | 9007298 | |
M74541.1 | - | 12 | 2472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
