siRNA Details
Gene Symbol: Cdh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007666.3 | NP_031692.2 | 9, 10 | 1771 | ||
GenBank mRNA | BC107008.1 | - | 8, 9 | 1512 | 40047565 | |
BC107013.1 | - | 8, 9 | 1512 | 40048308 | ||
D82029.1 | - | 8, 9 | 1512 | |||
U67399.1 | - | 1, 2 | 79 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
