siRNA Details
Gene Symbol: Cdk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007668.3 | NP_031694.1 | 3 | 217 | ||
GenBank mRNA | AK051790.1 | - | 3 | 217 | ||
BC052007.1 | - | 3 | 191 | 5706932 | ||
D29678.1 | - | 3 | 226 |

More Information
Other Species Targeted

Gene Ontology Categories:
