siRNA Details
Gene Symbol: Cdk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009873.2 | NP_034003.1 | 2 | 381 | ||
GenBank mRNA | AF132482.1 | - | 2 | 212 | ||
AF132483.1 | - | 2 | 381 | |||
AK078973.1 | - | 2 | 492 | |||
AK154197.1 | - | 3 | 654 | |||
BC119360.1 | - | 2 | 370 | 8734109 | ||
BC120595.1 | - | 2 | 370 | 40129518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
