siRNA Details
Gene Symbol: Cdk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009873.2 | NP_034003.1 | 4 | 691 | ||
GenBank mRNA | AF132482.1 | - | 4 | 522 | ||
AF132483.1 | - | 4 | 691 | |||
AK078973.1 | - | 4 | 802 | |||
AK154197.1 | - | 5 | 964 | |||
BC119360.1 | - | 4 | 680 | 8734109 | ||
BC120595.1 | - | 4 | 680 | 40129518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
