siRNA Details
Gene Symbol: Cdk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009874.3 | NP_034004.2 | 4 | 313 | ||
GenBank mRNA | AK039084.1 | - | 4 | 301 | ||
AK145759.1 | - | 4 | 239 | |||
AK168286.1 | - | 4 | 312 | |||
BC004605.1 | - | 4 | 271 | 3585145 | ||
BC068160.1 | - | 4 | 298 | 30358255 | ||
BC141043.1 | - | 4 | 270 | 9055655 | ||
CJ111227.1 | - | 4 | 315 | |||
U11822.1 | - | 4 | 276 | |||
X74145.1 | - | 4 | 295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
