siRNA Details
Gene Symbol: Cdkn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009875.4 | NP_034005.2 | 1 | 701 | ||
GenBank mRNA | AK046676.1 | - | 1 | 436 | ||
AK047669.1 | - | 1 | 702 | |||
AK050240.1 | - | 1 | 688 | |||
AK158411.1 | - | 1 | 681 | |||
AK162146.1 | - | 1 | 421 | |||
BC014296.1 | - | 1 | 684 | 4193493 | ||
CT010382.1 | - | 1 | 182 | |||
U09968.1 | - | 1 | 182 | |||
U10440.1 | - | 1 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
