siRNA Details
Gene Symbol: Cdkn2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009878.3 | NP_034008.2 | 2 | 489 | ||
GenBank mRNA | AK132906.1 | - | 2 | 454 | ||
AK140808.1 | - | 2 | 166 | |||
AK144737.1 | - | 3 | 487 | |||
BC013898.1 | - | 2 | 318 | 4020283 | ||
U19597.1 | - | 2 | 362 | |||
U20497.1 | - | 2 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
