siRNA Details
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA016357.1 | - | 2 | 234 | ||
AF012709.1 | - | 2 | 290 | |||
AK010718.1 | - | 2 | 252 | |||
AK011399.1 | - | 2 | 235 | |||
AK040313.1 | - | 3 | 417 | |||
AK041138.1 | - | 3 | 410 | |||
AK053763.1 | - | 2 | 259 | |||
AK089033.1 | - | 2 | 263 | |||
AK182390.1 | - | 2 | 196 | |||
BC011038.1 | - | 2 | 270 | 4018429 | ||
BC012280.1 | - | 4 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
