siRNA Details
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007694.4 | NP_031720.1 | 4 | 1810 | ||
GenBank mRNA | AK077487.1 | - | 4 | 1379 | ||
AK077489.1 | - | 4 | 1418 | |||
AK133629.1 | - | 4 | 1415 | |||
AK133739.1 | - | 4 | 1419 | |||
AK133786.1 | - | 4 | 1420 | |||
AK158857.1 | - | 4 | 1811 | |||
BC014736.1 | - | 4 | 1411 | 4511248 | ||
X51429.1 | - | 4 | 1406 | |||
X53028.1 | - | 4 | 1416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
