siRNA Details
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162410.1 | NP_001155882.1 | 2 | 217 | ||
NM_007700.2 | NP_031726.2 | 2 | 217 | |||
GenBank mRNA | AK018671.1 | - | 2 | 196 | ||
AK035326.1 | - | 2 | 174 | |||
AK150039.1 | - | 2 | 196 | |||
AK155328.1 | - | 2 | 197 | |||
AK155393.1 | - | 2 | 218 | |||
AK164236.1 | - | 2 | 218 | |||
AK165275.1 | - | 1 | 3728 | |||
BC018243.1 | - | 2 | 209 | 4511275 | ||
U12473.1 | - | 2 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
