siRNA Details
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162410.1 | NP_001155882.1 | 8 | 841 | ||
NM_007700.2 | NP_031726.2 | 8 | 841 | |||
GenBank mRNA | AK018671.1 | - | 8 | 820 | ||
AK035326.1 | - | 8 | 798 | |||
AK150039.1 | - | 8 | 820 | |||
AK155328.1 | - | 8 | 821 | |||
AK155393.1 | - | 8 | 842 | |||
AK164236.1 | - | 8 | 842 | |||
BC018243.1 | - | 8 | 833 | 4511275 | ||
U12473.1 | - | 8 | 809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
