siRNA Details
Gene Symbol: Cideb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009894.3 | NP_034024.2 | 4 | 620 | ||
GenBank mRNA | AF041377.1 | - | 4 | 608 | ||
AK003968.1 | - | 4 | 656 | |||
BC012664.1 | - | 4 | 584 | 4218866 | ||
BC046340.1 | - | 4 | 614 | 6478230 | ||
CB600615.1 | - | 4 | 597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
