siRNA Details
Gene Symbol: Ckb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021273.4 | NP_067248.1 | 5, 6 | 786 | ||
GenBank mRNA | AK002467.1 | - | 5, 6 | 749 | ||
AK014299.1 | - | 5, 6 | 729 | |||
AK148885.1 | - | 5, 6 | 746 | |||
AK152388.1 | - | 5, 6 | 746 | |||
AK153484.1 | - | 5, 6 | 746 | |||
AK161990.1 | - | 5, 6 | 746 | |||
AK166980.1 | - | 5, 6 | 749 | |||
AK167034.1 | - | 5, 6 | 749 | |||
BC015271.1 | - | 5, 6 | 716 | 4225384 | ||
BC106109.1 | - | 5, 6 | 713 | 6395794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
