siRNA Details
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007711.3 | NP_031737.1 | 11 | 2733 | ||
NM_173873.1 | NP_776298.1 | 12 | 2909 | |||
NM_173876.3 | NP_776301.1 | 11 | 2454 | |||
GenBank mRNA | AF029347.1 | - | 11 | 2357 | ||
AK141136.1 | - | 12 | 2847 | |||
AK147364.1 | - | 10 | 6932 | |||
AK147548.1 | - | 10 | 6934 | |||
AK147621.1 | - | 10 | 6934 | |||
AK159698.1 | - | 12 | 2883 | |||
AK161327.1 | - | 12 | 2837 | |||
AK162639.1 | - | 12 | 2850 | |||
BC057133.1 | - | 11 | 2387 | 6833732 | ||
X78874.1 | - | 11 | 2558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
