siRNA Details
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007711.3 | NP_031737.1 | 8 | 1826 | ||
NM_173873.1 | NP_776298.1 | 9 | 2002 | |||
NM_173874.1 | NP_776299.1 | 9 | 2002 | |||
NM_173876.3 | NP_776301.1 | 8 | 1547 | |||
GenBank mRNA | AF029347.1 | - | 8 | 1450 | ||
AK141136.1 | - | 9 | 1940 | |||
AK147364.1 | - | 8 | 1982 | |||
AK147548.1 | - | 8 | 1984 | |||
AK147621.1 | - | 8 | 1984 | |||
AK149276.1 | - | 9 | 1541 | |||
AK150688.1 | - | 9 | 1931 | |||
AK159698.1 | - | 9 | 1976 | |||
AK161327.1 | - | 9 | 1930 | |||
AK162639.1 | - | 9 | 1943 | |||
AK169297.1 | - | 9 | 1923 | |||
AK169516.1 | - | 3 | 1878 | |||
BC057133.1 | - | 8 | 1480 | 6833732 | ||
BC057855.1 | - | 8 | 1642 | |||
X78874.1 | - | 8 | 1651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
