siRNA Details
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007711.3 | NP_031737.1 | 10 | 2288 | ||
NM_173873.1 | NP_776298.1 | 11 | 2464 | |||
NM_173874.1 | NP_776299.1 | 11 | 2464 | |||
NM_173876.3 | NP_776301.1 | 10 | 2009 | |||
GenBank mRNA | AF029347.1 | - | 10 | 1912 | ||
AK141136.1 | - | 11 | 2402 | |||
AK147364.1 | - | 9 | 6487 | |||
AK147548.1 | - | 9 | 6489 | |||
AK147621.1 | - | 9 | 6489 | |||
AK149276.1 | - | 11 | 2003 | |||
AK150688.1 | - | 11 | 2393 | |||
AK159698.1 | - | 11 | 2438 | |||
AK161327.1 | - | 11 | 2392 | |||
AK162639.1 | - | 11 | 2405 | |||
AK169297.1 | - | 11 | 2385 | |||
BC057133.1 | - | 10 | 1942 | 6833732 | ||
BC057855.1 | - | 10 | 2104 | |||
X78874.1 | - | 10 | 2113 |

More Information
Other Species Targeted

Gene Ontology Categories:
