siRNA Details
Gene Symbol: Clcnka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF124848.1 | - | 11 | 1246 | ||
BC037077.1 | - | 12 | 1157 | 5042736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
