siRNA Details
Gene Symbol: Cldn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016675.4 | NP_057884.1 | 2 | 579 | ||
GenBank mRNA | AF072128.1 | - | 1 | 241 | ||
AK004990.1 | - | 1 | 346 | |||
BC015252.1 | - | 1 | 319 | 4223446 | ||
BC085494.1 | - | 2 | 559 | 5686202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
