siRNA Details
Gene Symbol: Cldn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013805.4 | NP_038833.2 | 1 | 1069 | ||
GenBank mRNA | AF035814.1 | - | 1 | 1061 | ||
AK077282.1 | - | 1 | 1070 | |||
BC002016.1 | - | 1 | 1053 | 3488769 | ||
BC083341.1 | - | 1 | 1059 | 6819173 | ||
U82758.1 | - | 1 | 942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
