siRNA Details
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042634.2 | NP_001036099.1 | 2, 3 | 333 | ||
GenBank mRNA | AK031218.1 | - | 2, 3 | 267 | ||
AK076199.1 | - | 2, 3 | 268 | |||
AK135765.1 | - | 2, 3 | 268 | |||
BC062887.1 | - | 285 | ||||
BC145905.1 | - | 2, 3 | 236 | |||
BC145911.1 | - | 2, 3 | 236 | |||
L29221.1 | - | 2, 3 | 249 | |||
M38381.1 | - | 2, 3 | 249 | |||
U11054.1 | - | 2, 3 | 233 | |||
U21209.1 | - | 2, 3 | 249 | |||
X57186.1 | - | 2, 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
