siRNA Details
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013492.2 | NP_038520.2 | 5 | 884 | ||
GenBank mRNA | AF248057.1 | - | 5 | 767 | ||
AF248058.1 | - | 2 | 438 | |||
BC075668.1 | - | 5 | 713 | 30550773 | ||
D14077.1 | - | 5 | 742 | |||
L05670.1 | - | 5 | 742 | |||
L08235.1 | - | 4 | 714 | |||
S70244.1 | - | 4 | 678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
