siRNA Details
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013492.2 | NP_038520.2 | 8 | 1505 | ||
GenBank mRNA | AF248057.1 | - | 8 | 1388 | ||
AF248058.1 | - | 5 | 1059 | |||
BC075668.1 | - | 8 | 1334 | 30550773 | ||
CK623407.1 | - | 2 | 160 | |||
D14077.1 | - | 8 | 1363 | |||
L05670.1 | - | 8 | 1363 | |||
L08235.1 | - | 7 | 1335 | |||
S70244.1 | - | 7 | 1299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
