siRNA Details
Gene Symbol: Ccr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007720.2 | NP_031746.1 | 2 | 650 | ||
GenBank mRNA | BC101966.1 | - | 1 | 594 | 40044931 | |
BC101967.1 | - | 1 | 594 | 40044932 | ||
BC103566.1 | - | 1 | 594 | 40044933 | ||
BC103567.1 | - | 1 | 594 | 40044935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
