siRNA Details
Gene Symbol: Cnih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009919.2 | NP_034049.2 | 4 | 465 | ||
GenBank mRNA | AF022811.1 | - | 4 | 456 | ||
AK002293.1 | - | 4 | 433 | |||
AK011200.1 | - | 4 | 418 | |||
AK152432.1 | - | 4 | 416 | |||
AK154479.1 | - | 4 | 416 | |||
AK168154.1 | - | 4 | 416 | |||
BC034868.1 | - | 4 | 425 | 2938052 | ||
DV653213.1 | - | 4 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
