siRNA Details
Gene Symbol: Cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB006191.2 | - | 6 | 1123 | ||
AK141483.1 | - | 6 | 1223 | |||
BC059922.1 | - | 6 | 1209 | |||
BC103797.1 | - | 6 | 1197 | 30134114 | ||
CX243776.1 | - | 1 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
