siRNA Details
Gene Symbol: Cnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007725.2 | NP_031751.1 | 5 | 513 | ||
GenBank mRNA | AK051845.1 | - | 5 | 514 | ||
AK189304.1 | - | 5 | 441 | |||
BC008181.1 | - | 5 | 431 | 3589470 | ||
BC009144.1 | - | 5 | 468 | 3594856 | ||
BC018482.1 | - | 5 | 483 | 3589158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
