siRNA Details
Gene Symbol: Col2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031163.3 | NP_112440.2 | 2 | 351 | ||
GenBank mRNA | BC082331.1 | - | 2 | 284 | 30537724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
