siRNA Details
Gene Symbol: Col4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009932.3 | NP_034062.3 | 47 | 5174 | ||
GenBank mRNA | AY375463.1 | - | 2 | 403 | ||
AY502947.1 | - | 1 | 118 | |||
BC013560.1 | - | 5 | 831 | |||
BC138040.1 | - | 46 | 4886 | 8861059 | ||
J04695.1 | - | 47 | 4979 | |||
M15833.1 | - | 2 | 403 | |||
X04647.1 | - | 15 | 1936 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008771417.
Gene Ontology Categories:
