siRNA Details
Gene Symbol: Col5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015734.2 | NP_056549.2 | 4 | 932 | ||
GenBank mRNA | AB009993.1 | - | 4 | 672 | ||
BC138447.1 | - | 4 | 606 | 8861467 | ||
BC138449.1 | - | 4 | 606 | 8861469 | ||
S63661.1 | - | 3 | 436 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_134452, XM_006233873, AF272662.
Gene Ontology Categories:
