siRNA Details
Gene Symbol: Col9a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009936.2 | NP_034066.2 | 32 | 2503 | ||
GenBank mRNA | AF237721.1 | - | 7 | 1157 | ||
AK003258.1 | - | 4 | 891 | |||
AK030350.1 | - | 32 | 2482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
